Orders LinkContactUsLinkComponent
Halogen lamp
Halogen lamp
Narva halogen lamp, 6 V 25 W, with base PY16-1,25, luminous flux 510 lm, life 100 h, filament of 1,7×1,7 mm, Medical and scientific applications, 25 W
Catalog # TSSP55147
Halogen lamp
Catalog # TSSP55147
Halogen lamp
Halogen lamp
Narva halogen lamp, 6 V 25 W, with base PY16-1,25, luminous flux 510 lm, life 100 h, filament of 1,7×1,7 mm, Medical and scientific applications, 25 W
Price
122,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Narva halogen special lamps are specifically designed for special lighting tasks. These lamps with prefocus base are suitable for scientific-technical equipment due to their design and optical precision.
  • Made of quartz glass
  • Transmits UV radiation
  • Operating position of s90
  • With prefocus base and mounting plate
Applications: Microscopy, ophthalmology, dermatology, food analysis, blood analysis, illumination for surgeries and analysis, measurement technology, projection.
Specifications
  • For:
    Medical and scientific applications
  • Description:
    Narva halogen lamp, 6 V 25 W, with base PY16-1,25, luminous flux 510 lm, life 100 h, filament of 1,7×1,7 mm
  • Power:
    25 W