ESD occupational shoes, lace-up, Uni6, 36750/36751
Lieferant: Abeba
Zertifikate
Über diesen Artikel
ESD shoes made from smooth leather, reflective material and a TPU sole resistant to oil, fuel and animal fats.
- Perspiration-absorbent special lining with Silver Point
- Replaceable acc Wave insole; orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- PU midsole
- Slip resistant TPU sole
Suitable for use in kitchens, food processing industries.
Colour: White, black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O2, FO, SRC.
Frequently Bought Together