Order Entry
Austria
ContactUsLinkComponent
VWR® MCP 100MCT 100 Pocket Colorimeters
VWR® MCP 100MCT 100 Pocket Colorimeters
Supplier:  VWR Collection
Certificates
undefined
VWR® MCP 100MCT 100 Pocket Colorimeters
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 705-1085
  • 705-1084
  • Model:
  • MCT 100
  • MCP 100
  • Optics:
  • LED, interference filter (IF) and photo sensor in transparent sample chamber
  • LED, interference filter (IF) and photo sensor in transparent sample chamber
  • Range:
  • 0,01 - 10 mg/L
  • 0,02 - 8 mg/L
  • Wavelength accuracy:
  • ±1 nm
  • ±1 nm
  • Display:
  • Backlit LCD (on key press)
  • Backlit LCD (on key press)
  • Power supply:
  • 4 micro batteries (AAA), capacity approx. 17 h or 5000 tests
  • 4 micro batteries (AAA), capacity approx. 17 h or 5000 tests
  • Automatic switch-off:
  • Yes
  • Yes
  • W×D×H:
  • 75×155×35 mm
  • 75×155×35 mm
  • Weight:
  • 260 g
  • 260 g
  • Absorbance:
  • –0,1...2,5 Abs
  • –0,1...2,5 Abs
  • Sensor:
  • Silicium photocell
  • Silicium photocell

Specifications

Related Information

Reference standard kits

Product Information