Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 1 mol/l (2 N) in aqueous solution for biopharmaceutical production
Sulphuric acid 1 mol/l (2 N) in aqueous solution for biopharmaceutical production
Catalog # 6501.2500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid 1 mol/l (2 N) in aqueous solution for biopharmaceutical production
Catalog # 6501.2500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer:
    0.996 --> 1.004 mol/l
  • Appearance:
    Passes test
  • Identification:
    Passes test
  • Residual metal catalyst:
    Unlikely to be present
  • Residual solvents (ICH Q3C):
    Excl.by manufact. process
  • Endotoxin:
    Max. 2.5 EU/ml
  • Filtered through:
    0.2 μm Confirmed

Specification Test Results

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:3264
ADR:8,III
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589