Wavelength calibration solution for ICP-OES and MP-AES
Catalog # VARI6610030100
Supplier: VARIAN
CAS Number:
Specifications
- Pk:1
- Description:Wavelength calibration solution, ready to use, for ICP-OES and MP-AES, 500 ml
Specifications
About this item
500 ml solution contains 5 mg/l Al, As, Ba,Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn and 50 mg/l K in 5 % HNO3 + tr HF.
- Ready to use
- Packaged in pre-leached HDPE bottles, secured with a tamper-evident seal
Delivery information: Supplied with certificate of analysis.