Orders LinkContactUsLinkComponent
Objectives, Panthera
 :   MOTIC
Objectives, Panthera
 :  MOTIC
Objectives, Panthera
Objectives, Panthera
Product Family Options
  • Description
    Magnification
    Working distance
  • CCIS® Plan achromatic objective UC PL 2×/0,05 (WD=7,2 mm) (To be used with swing-out condenser)
    2x/0,05
    7,2 mm
    1 * 1 ST
    1 * 1 ST - 279,00 €
    279,00 €
    1 * 1 ST
       279,00 €
  • CCIS® Plan achromatic objective UC PL 4×/0,1 (WD=30,5 mm)
    4x/0,1
    30,5 mm
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • CCIS® Plan achromatic objective UC PL 10×/0,25 (WD=17,4 mm)
    10x/0,25
    17,4 mm
    1 * 1 ST
    1 * 1 ST - 67,00 €
    67,00 €
    1 * 1 ST
       67,00 €
  • CCIS® Plan achromatic objective UC PL 20×/0,45 (WD=0,8 mm)
    20x/0,45
    0,8 mm
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • CCIS® Plan achromatic objective UC PL 40×/0,65/S (WD=0,6 mm)
    40x/0,65
    0,6 mm
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • CCIS® Plan achromatic objective UC PL 60×/0,8/S (WD=0,35 mm)
    60x/0,8
    0,35 mm
    1 * 1 ST
    1 * 1 ST - 167,00 €
    167,00 €
    1 * 1 ST
       167,00 €
  • CCIS® Plan achromatic objective UC PL 100×/1,25/S-Oil (WD=0,16 mm)
    100x/1,25 (i)
    0,16 mm
    1 * 1 ST
    1 * 1 ST - 173,00 €
    173,00 €
    1 * 1 ST
       173,00 €
  • CCIS® Plan achromatic objective UC PL 100×/0,8/S-Dry (WD=1,7 mm)
    100x/0,8
    1,7 mm
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €
  • CCIS® Plan achromatic Phase objective UC PL Ph 10×/0,25 (WD=17,4 mm), Ph1
    Ph1 10x/0,25
    17,4 mm
    1 * 1 ST
    1 * 1 ST - 206,00 €
    206,00 €
    1 * 1 ST
       206,00 €
  • CCIS® Plan achromatic Phase objective UC PL Ph 20×/0,45 (WD=0,8 mm), Ph2
    Ph2 20x/0,45
    0,8 mm
    1 * 1 ST
    1 * 1 ST - 252,00 €
    252,00 €
    1 * 1 ST
       252,00 €
  • CCIS® Plan achromatic Phase objective UC PL Ph 40×/0,65/S (WD=0,6 mm), Ph2
    Ph2 40x/0,65
    0,6 mm
    1 * 1 ST
    1 * 1 ST - 390,00 €
    390,00 €
    1 * 1 ST
       390,00 €
  • CCIS® Plan achromatic Phase objective UC PL Ph 100×/1,25/S-Oil (WD=0,16 mm), Ph3
    Ph3 100x/1,25 (i)
    0,16 mm
    1 * 1 ST
    1 * 1 ST - 567,00 €
    567,00 €
    1 * 1 ST
       567,00 €
  • CCIS® Plan achromatic objective UC PL NCG 10×/0,25 (WD=17,4 mm)
    10x/0,25
    17,4 mm
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • CCIS® Plan achromatic objective UC PL NCG 20×/0,45 (WD=0,8 mm)
    20x/0,45
    0,8 mm
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
  • CCIS® Plan achromatic objective UC PL NCG 40×/0,65/S (WD=0,6 mm)
    40x/0,65
    0,6 mm
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €
  • CCIS® Plan achromatic objective UC PL NCG 100×/1,25/S-Oil (WD=0,16 mm)
    100x/1,25 (i)
    0,16 mm
    1 * 1 ST
    1 * 1 ST - 203,00 €
    203,00 €
    1 * 1 ST
       203,00 €