VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
Cardboard, coated.
- Waterproof coating
- Freezable to –140 °C
- Available with or without dividers
- In different heights and five colours
For storage of cryovials at low temperatures.
Zugehörige Informationen
Frequently Bought Together