Express Eingabe
Austria
ContactUsLinkComponent
Single element petroleum standard, sulphur free lead in hydrocarbon oil, CRM, VHG
Single element petroleum standard, sulphur free lead in hydrocarbon oil, CRM, VHG
Catalog # PROOVHG-OSF-PB1000
CAS Number:  
Single element petroleum standard, sulphur free lead in hydrocarbon oil, CRM, VHG
Catalog # PROOVHG-OSF-PB1000
CAS Number:  

Specifications

  • Konzentration:
    1000 µg/g
  • VE:
    50 g
  • Bezeichnung:
    Sulphur-free standard
  • Element:
    Blei
  • Symbol:
    Pb

Specifications

About this item

This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or other organic matrices using x-ray fluorescence (XRF) spectrometry, inductively coupled plasma optical emission spectroscopy (ICPOES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.

Certifications: Traceable to NIST, SRM.