Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Supplier: LGC Standards PROMOCHEM
Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Supplier: LGC Standards PROMOCHEM
Certificates
Jump to:
About this item
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
: Traceable to NIST, SRM.
4 Options Available Below
Product Details & Documents
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
: Traceable to NIST, SRM.
Product Family Options
Product Information
- MatrixPkAvailabilityPrice
- PROOVHG-MMGN1P-1001% Magnesium nitrate, 100 ml
- PROOVHG-MPM1-250Pre-mixed, 750 µg/ml Pd and 500 µg/ml Mg(NO₃)₂, 250 mlPre-mixed, 750 µg/ml Pd and 500 µg/ml Mg(NO₃)₂, 250 ml2% HNO₃250 mLLog in for availability1 * 250 mL88,90 €
- PROOVHG-MPM2-250Pre-mixed, 1000 µg/ml Pd and 600 µg/ml Mg(NO₃)₂, 250 mlPre-mixed, 1000 µg/ml Pd and 600 µg/ml Mg(NO₃)₂, 250 ml2% HNO₃250 mLLog in for availability1 * 250 mL105,00 €
- PROOVHG-MPM3-250Pre-mixed, 10 mg/ml NH₄H₂PO₄ and 500 µg/ml Mg(NO₃)₂, 250 mlPre-mixed, 10 mg/ml NH₄H₂PO₄ and 500 µg/ml Mg(NO₃)₂, 250 ml2% HNO₃250 mLLog in for availability1 * 250 mL72,80 €