ContactUsLinkComponent
Flask, Digesdahl®
Flask, Digesdahl®
Catalog # 458-0048
Supplier:  Hach
undefined
Flask, Digesdahl®
Catalog # 458-0048
Supplier:  Hach
Specifications
  • Description:
    Flask, flat-bottom
  • Pk:
    1
Specifications
About this item
Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.