Standards for water pollution QC check, total phenolics, VHG
Catalog # PROOVHG-QWPPHEN-2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:2 mL
- Description:Water analysis standard for total phenolics
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to total phenolics.
This standard has been prepared in methanol as a concentrate and must be diluted prior to analysis. This concentrate is not preserved.
Certifications: Traceable to NIST SRM.