ContactUsLinkComponent
Microtube membranes, breathable, Breathe-EASIER
Microtube membranes, breathable, Breathe-EASIER
Catalog # DIVEBMTM-2000
Microtube membranes, breathable, Breathe-EASIER
Catalog # DIVEBMTM-2000
Specifications
  • Description:
    Microtube membranes
  • Pk:
    100
Specifications
About this item
Breathe-EASIER micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange. Formatted with an easy to use finger tab, membranes are precut to fit a variety of tube sizes. Membranes are gamma irradiated to ensure sterility and are free of cytotoxins.
  • Microtube cap closures will not bind in centrifuge rotors
  • Convenient scoring makes liner removal quick and easy
  • White non woven rayon membrane rapidly transmits water vapour for accelerated growth