Order Entry
Austria
ContactUsLinkComponent
Cell culture media, Minimum Essential Medium (MEM), SAFC®
Cell culture media, Minimum Essential Medium (MEM), SAFC®
Catalog # SIAL51412C-1000ML
Supplier:  Merck
CAS Number:  
Cell culture media, Minimum Essential Medium (MEM), SAFC®
Catalog # SIAL51412C-1000ML
Supplier:  Merck
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    1.000 mL
  • Description:
    MEM with Earle′s balanced salts, without L-glutamine, sterile filtered

Specifications

About this item

Minimum Essential Medium (MEM) is a modification of Eagle’s earlier medium, Basal Medium Eagle (BME), containing higher concentrations of the essential nutrients. This medium has demonstrated the ability to support a variety of normal and transformed cells in culture and contains Earle’s balanced salts, which makes it suitable for use in atmospheres charged with CO₂ gas.

Storage temperature: 2 to 8 °C.