ContactUsLinkComponent
Beakers, low form, quartz, with spout
Beakers, low form, quartz, with spout
Catalog # TSLA75203
Supplier #: 75203
Beakers, low form, quartz, with spout
Catalog # TSLA75203
Supplier #: 75203
Log in for availability
Select an Option
44 mm
Select an Option
50 ml
Select an Option
55 mm
Select an Option
04
Select an Option
1
About this item
Temperature-resistant beakers made from transparent quartz glass.
  • High chemical resistance
  • High resistance against thermal schock
  • Made for tasks up to 1050 °C
  • High purity
  • Without graduations
Frequently Bought Together