Order Entry
Austria
ContactUsLinkComponent
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Catalog # SUPL39361-250ML
Supplier:  Merck
CAS Number:  
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Catalog # SUPL39361-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    2% HNO₃
  • Pack type:
    HDPE bottle
  • Pk:
    250 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item

This silver standard solution prepared with high quality silver metal in 2% HNO3.

Certifications: ISO/IEC 17025 and ISO Guide 34. Traceable to NIST, SRM.