Reference standards, PAH mixture 15
According to EU recommendation about the determination of carcinogenic PAHs in food (2005/108/EC) .
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
MP-AES torches
: Agilent