COD standard solutions, VHG
Catalog # PROOVHG-COD1K-100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/l
- Matrix:H₂O
- Pk:100 mL
- Description:COD, 0,5% H₂SO₄
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a calibration standard or quality control check for chemical oxygen demand (COD).
Certifications: Traceable to NIST SRM.