Order Entry
Austria
ContactUsLinkComponent
BOD standard solutions, VHG
BOD standard solutions, VHG
Catalog # PROOVHG-BOD200A
CAS Number:  
BOD standard solutions, VHG
Catalog # PROOVHG-BOD200A
CAS Number:  

Specifications

  • Concentration:
    200 mg/l
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    BOD standard
  • Seq:
    01
  • Type:
    Reference standard

Specifications

About this item

The standard solution is intended for use as a glucose-glutamic acid standard check solution for biochemical oxygen demand (BOD) in APHA Method 5210 B.

Certifications: Traceable to NIST.