Multi-element standards, common elements mix 2, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES), and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V and Zn.
Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Frequently Bought Together