Reference materials, cloud point reference, VHG
Cloud point range, +3 to +9 °C, 250 ml, 250 mL
Catalog # PROOVHG-C5-250
Supplier: LGC Standards PROMOCHEM
Reference materials, cloud point reference, VHG
Catalog # PROOVHG-C5-250
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
These reference materials are intended for use in accordance with ASTM D2500 for the determination of the cloud point of petroleum products. The cloud point of a petroleum product is an index of the lowest temperature of their utility for certain applications.
Lieferumfang: Each standard is supplied with a full certificate of analysis (COA) that states the certified value.
Achtung! Bitte beachten: Not intended for use with phase instruments.
Specifications
Specifications
- Pk:250 mL
- Description:Cloud point range, +3 to +9 °C, 250 ml



