Ammonium magnesium phosphate hydrate ≥99.997% (trace metals basis), Sigma-Aldrich®
SIAL529354-50G
: Merck
:
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
- Linear Formula:(NH₄)MgPO₄ · xH₂O
- Molecular Weight:137,31 (anhydrous basis)
- EC Number:232-075-2
- MDL Number:MFCD03427023
- PubChem Substance ID:24877674
- InChI Key:DRMWBNMSWMDKLG-UHFFFAOYSA-L
- Assay:99,997% trace metals basis
- Form:Solid
- Synonym:Phosphoric acid ammonium magnesium salt hydrate
:
:137.32 g/mol (anhy)
:7785-21-9
:NH₄MgPO₄·xH₂O
:MFCD00050358
:M-Clarity™ quality level = MQ100