Order Entry
Austria
ContactUsLinkComponent
VWR®, Secondary reference standards for volumetric titration
VWR®, Secondary reference standards for volumetric titration
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Secondary reference standards for volumetric titration
Supplier:  VWR Chemicals
Certificates

Specifications

  • CAS No.:
    7778-50-9
  • Formula:
    K₂Cr₂O₇
  • Description:
    Potassium dichromate

Specifications

Related Information

Frequently Bought Together