Order Entry
Austria
ContactUsLinkComponent
Colour 500 Pt-Co units calibration standard, CRM
Colour 500 Pt-Co units calibration standard, CRM
Catalog # PROORTCCLR500100ML
CAS Number:  
Colour 500 Pt-Co units calibration standard, CRM
Catalog # PROORTCCLR500100ML
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Pt-Co, 500 units colour

Specifications

About this item

Certifications: Traceable NIST, SRM.