


Tetrahydrofuran ≥99.8% (by GC, corrected for water content) contains no preservatives, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay (by GC):min. 99.8 %
- Peroxide (as H2O2):max. 0.010 %
- Residue after Evaporation:max. 5 ppm
- Water (H2O):max. 0.02 %
- at 230 nm:max. 1.0
- at 260 nm:max. 0.15
- at 270 nm:max. 0.08
- at 310 nm:max. 0.02
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR9441
SDS
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Frequently Bought Together