Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Carbonate Standards
Calcium carbonate, alkalinity standard, CRM
Calcium carbonate, alkalinity standard, CRM
Calcium carbonate, alkalinity standard, CRM
Catalog # PROORTCALK1000-500
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Calcium carbonate, alkalinity standard, CRM
Catalog # PROORTCALK1000-500
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Concentration:
1000 mg/l
Matrix:
H₂O
Pk:
500 mL
Description:
Calcium carbonate alkalinity standard
Seq:
02
Specifications
About this item
Product Details
Product Details