Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standards, β-zearalenol (NMIA P1796)
Catalog # PROONMIAP1796
CAS Number:  
Organic reference standards, β-zearalenol (NMIA P1796)
Catalog # PROONMIAP1796
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Reference standard
  • CAS No.:
    71030-11-0
  • Description:
    β-Zearalenol
  • Formula:
    C₁₈H₂₄O₅
  • Description:
    β-Zearalenol

Specifications

About this item

The purity estimate by traditional analytical techniques was obtained by subtraction from 100% of total impurities by HPLC with UV detection, Karl Fischer analysis, thermogravimetric analysis and 1H NMR is 99,3 ± 1,3% mass fraction, (95% coverage interval). Supporting evidence is provided by elemental microanalysis.

  • White crystalline powder
  • Moisture content ≤0,7% mass fraction
  • Volatile content and non volatile residue ≤0,3% mass fraction
  • CAS no.: 71030-11-0

Synonyms: (3S,7S,11E)-3,4,5,6,7,8,9,10-octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one.