Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, 16β-hydroxystanozolol (NMIA D621), Certified Reference Material (CRM)
Organic reference standards, 16β-hydroxystanozolol (NMIA D621), Certified Reference Material (CRM)
  PROONMIAD621
 :  
Organic reference standards, 16β-hydroxystanozolol (NMIA D621), Certified Reference Material (CRM)
  PROONMIAD621
 :  

 

  • Pk:
    1 mg
  • Standards:
    Reference standard
  • CAS No.:
    125590-76-3
  • Description:
    16β-Hydroxystanozolol
  • Formula:
    C₂₁H₃₄N₂O₂
  • Description:
    16β-Hydroxystanozolol

 

 

Purity estimate obtained by quantitative NMR against an internal standard of dimethyl sulphone is 84,6 ± 1,5% mass fraction, (95% coverage interval). Supporting evidence is provided by HPLC with UV detection, Karl Fischer analysis and theromogravimetric analysis.

  • White crystals
  • Metabolite of stanozolol
  • Moisture content ≤0,6% mass fraction
  • CAS no.: 125590-76-3

Synonyms: 16β,17β-dihydroxy-17α-methyl-5α-androstano[3,2-c]pyrazole.