ContactUsLinkComponent
Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
 :  Avantor
Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
 :  Avantor
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Particulate Matter:
    max. 0.0001 %
  • Residue after Evaporation:
    max. 2 ppm
  • at 220 nm:
    max. 0.005 au
  • at 254 nm:
    max. 0.001 au
Frequently Bought Together