Sodium dihydrogen phosphate monohydrate, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208
:137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:0.9996 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Assay:98.0 - 102.0 %
- Calcium (Ca):max. 0.005 %
- Iron (Fe):max. 0.001 %
- Potassium (K):max. 0.01 %
- Heavy Metals (as Pb):max. 0.001 %
- Insoluble Matter:max. 0.01 %
- Sulfate (SO4):max. 0.003 %
- Chloride (Cl):max. 5
Specification Test Results
Frequently Bought Together