Circulating water baths, Precision™
Precision™ CIR 35
HAKATSCIR35
These circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in serological applications.
- Enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Easily operate and monitor with icon-based graphical display
- Prevent bath overheating and damage with low-fluid protection
- Monochrome LCD display
: Supplied with stainless steel gable cover and rubber duck.
- Description:Precision™ CIR 35
- Cat. No.:HAKATSCIR35
- Temperature uniformity:±0,05 °C
- Capacity:35 L
- Heating capacity:1500 W
- W×D×H:394×938×249 mm
- Temperature range:RT +5...100 °C
- Electrical:200/230 V, 50/60 Hz
- Temperature stability:±0,1 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



