Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Silicate Standards
Silica standard solution, VHG
Silica standard solution, VHG
Silica standard solution, VHG
Catalog # PROOVHG-ISIO21K100
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Certificates
Expand image
Silica standard solution, VHG
Catalog # PROOVHG-ISIO21K100
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Certificates
Specifications
Concentration:
1000 mg/l
Matrix:
H₂O
Pk:
100 mL
Description:
Silica standard, SiO₂
Specifications
About this item
Product Details
Documentation
Product Details
Documentation