Order Entry
Austria
ContactUsLinkComponent
Entecavir Related Compounds, Reference Standard
Entecavir Related Compounds, Reference Standard
Supplier:  USP
Certificates
Entecavir Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    209216-23-9
  • Formula:
    C₁₂H₁₇N₅O₄
  • Description:
    Entecavir Monohydrate
  • Synonym(s):
    9-[(1S,3R,4S)-4-Dimethylphenylsilyl-3-(benzyloxymethyl)-2-methylenecyclopentyl]guanine

Specifications

Frequently Bought Together