ContactUsLinkComponent
Viscosity test standard for PTP, VHG
Viscosity test standard for PTP, VHG
  PROOG-VPTPVISC-50
 :  
Viscosity test standard for PTP, VHG
  PROOG-VPTPVISC-50
 :  
  • Pk:
    1
  • Description:
    Viscosity test standard, 40 °C and 100 °C
This proficiency testing program (PTP) was specially designed to give analytical laboratories instant validation while measuring viscosity in new and used oils.
 : Traceable to CRM. ISO/IEC 17043 accredited.