Order Entry
Austria
ContactUsLinkComponent
n-Hexane ≥95.0%, BAKER ANALYZED® HPLC, J.T.Baker®
n-Hexane ≥95.0%, BAKER ANALYZED® HPLC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
n-Hexane ≥95.0%, BAKER ANALYZED® HPLC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 95 %
  • Residue after Evaporation:
    max. 2 ppm
  • Substances Darkened by H2SO4:
    passes test
  • Water (H2O):
    max. 0.01 %
  • at 210 nm:
    max. 0.2
  • at 220 nm:
    max. 0.08
  • at 254 nm:
    max. 0.008
  • at 280 nm:
    max. 0.005
  • at 350 nm:
    max. 0.005
  • UV Cut-off, nm:
    max. 192

Specification Test Results

Related Information

Frequently Bought Together