Order Entry
Austria
ContactUsLinkComponent
n-Heptane, anhydrous (max. 0.003% H₂O)
Flammable
Health
Irritant
Environment
n-Heptane, anhydrous (max. 0.003% H₂O)
Catalog # 85818.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Heptane, anhydrous (max. 0.003% H₂O)
Catalog # 85818.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle with septum cap
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Assay (GC, on anhydrous basis):
    99.0-100 %
  • Acidity:
    0-0.001 meq/g
  • Residue after evaporation:
    0-0.001 % w/w
  • Water (KF):
    0-0.003 % w/w

Specification Test Results

About this item

 :
 :100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90,6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544