Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Prednisone, Ph.Eur., RS
Organic reference standard, Prednisone, Ph.Eur., RS
Catalog # EDQMP2900000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Prednisone, Ph.Eur., RS
Catalog # EDQMP2900000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    53-03-2
  • Description:
    Prednisone
  • Formula:
    C₂₁H₂₆O₅
  • Storage temperature:
    Ambient
  • Description:
    Prednisone

Specifications

About this item

  • In Monograph 0354
  • Solubility in water: less than 0,1