Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Paroxetine hydrochloride hemihydrate, Ph.Eur., RS
Organic reference standard, Paroxetine hydrochloride hemihydrate, Ph.Eur., RS
Catalog # EDQMY0000281
Supplier:  EDQM
CAS Number:  
Organic reference standard, Paroxetine hydrochloride hemihydrate, Ph.Eur., RS
Catalog # EDQMY0000281
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    Reference standard
  • CAS No.:
    110429-35-1
  • Description:
    Paroxetine hydrochloride hemihydrate
  • Formula:
    C₃₈H₄₄Cl₂F₂N₂O₇
  • Storage temperature:
    Ambient
  • Description:
    Paroxetine hydrochloride hemihydrate

Specifications

About this item

  • In Monographs 2018 and 2283