Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Nortriptyline hydrochloride, Ph.Eur., RS
Organic reference standard, Nortriptyline hydrochloride, Ph.Eur., RS
Catalog # EDQMN1280000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Nortriptyline hydrochloride, Ph.Eur., RS
Catalog # EDQMN1280000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    894-71-3
  • Description:
    Nortriptyline hydrochloride
  • Formula:
    C₁₉H₂₂ClN
  • Storage temperature:
    Ambient
  • Description:
    Nortriptyline hydrochloride

Specifications

About this item

  • In Monograph 0941