Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Maltitol, Ph.Eur., RS
Organic reference standard, Maltitol, Ph.Eur., RS
Catalog # EDQMM0160000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Maltitol, Ph.Eur., RS
Catalog # EDQMM0160000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1.200 mg
  • Standards:
    Reference standard
  • CAS No.:
    585-88-6
  • Description:
    Maltitol
  • Formula:
    C₁₂H₂₄O₁₁
  • Storage temperature:
    Ambient
  • Description:
    Maltitol

Specifications

About this item

  • In Monographs 1235 and 1236
  • Solubility in water
  • Pharmaceutical applications