Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Lincomycin hydrochloride, Ph.Eur., RS
Organic reference standard, Lincomycin hydrochloride, Ph.Eur., RS
Catalog # EDQML0650000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Lincomycin hydrochloride, Ph.Eur., RS
Catalog # EDQML0650000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    7179-49-9
  • Description:
    Lincomycin hydrochloride
  • Formula:
    C₁₈H₃₇ClN₂O₇S
  • Storage temperature:
    Ambient
  • Description:
    Lincomycin hydrochloride

Specifications

About this item

  • In Monographs 0582, 0583, and 0996
  • Soluble in/miscible with methanol