Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Kanamycin B sulfate, Ph.Eur., RS
Organic reference standard, Kanamycin B sulfate, Ph.Eur., RS
Catalog # EDQMK0100000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Kanamycin B sulfate, Ph.Eur., RS
Catalog # EDQMK0100000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    Reference standard
  • CAS No.:
    29701-07-3
  • Description:
    Kanamycin B sulfate
  • Formula:
    C₁₈H₃₉N₅O₁₄S
  • Storage temperature:
    Ambient
  • Description:
    Kanamycin B sulphate

Specifications

About this item

  • In Monographs 0032, 0033, and 0645
  • Solubility in water: 50 g/l at 20 °C
  • Pharmaceutical applications