Order Entry
Austria
ContactUsLinkComponent
Organic reference standards Fluoxetine, Ph.Eur., RS
Organic reference standards Fluoxetine, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards Fluoxetine, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    56296-78-7
  • Formula:
    C₁₇H₁₉ClF₃NO
  • Storage temperature:
    Ambient temperature
  • Description:
    Fluoxetine hydrochloride

Specifications

Frequently Bought Together