Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Ethinylestradiol, Ph.Eur., RS
Organic reference standard, Ethinylestradiol, Ph.Eur., RS
Catalog # EDQME1900000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Ethinylestradiol, Ph.Eur., RS
Catalog # EDQME1900000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    Reference standard
  • CAS No.:
    57-63-6
  • Description:
    Ethinylestradiol
  • Formula:
    C₂₀H₂₄O₂
  • Storage temperature:
    2 to 8°C
  • Description:
    Ethinylestradiol

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 0140, 0821, 0940, and 0926