Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, DL-Methionine, Ph.Eur., RS
Organic reference standard, DL-Methionine, Ph.Eur., RS
Catalog # EDQMM0950000
Supplier:  EDQM
CAS Number:  
Organic reference standard, DL-Methionine, Ph.Eur., RS
Catalog # EDQMM0950000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    59-51-8
  • Description:
    DL-Methionine
  • Formula:
    C₅H₁₁NO₂S
  • Storage temperature:
    Ambient
  • Description:
    DL-Methionine

Specifications

About this item

  • In Monograph 0624
  • Solubility in water: 34 g/l at 25°C