Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Dexamethasone sodium phosphate, Ph.Eur., RS
Organic reference standard, Dexamethasone sodium phosphate, Ph.Eur., RS
  EDQMD0720000
 :  EDQM
 :  
Organic reference standard, Dexamethasone sodium phosphate, Ph.Eur., RS
  EDQMD0720000
 :  EDQM
 :  

 

  • Pack type:
    Vial
  • Pk:
    130 mg
  • Standards:
    Reference standard
  • CAS No.:
    2392-39-4
  • Description:
    Dexamethasone sodium phosphate
  • Formula:
    C₂₂H₂₈FNa₂O₈P
  • Storage temperature:
    Ambient
  • Description:
    Dexamethasone sodium phosphate

 

 

Filled under inert atmosphere protected from humidity.

  • In Monographs 0549, 0735, and 0810
  • Soluble in water