Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Desmopressin, Ph.Eur., RS
Organic reference standard, Desmopressin, Ph.Eur., RS
Catalog # EDQMD0650000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Desmopressin, Ph.Eur., RS
Catalog # EDQMD0650000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    0,94 mg
  • Standards:
    Reference standard
  • CAS No.:
    62288-83-9
  • Description:
    Desmopressin
  • Formula:
    C₄₈H₆₈N₁₄O₁₄S₂
  • Storage temperature:
    -20°C ± 5°C
  • Description:
    Desmopressin

Specifications

About this item

  • In Monograph 0712
  • Soluble in water