Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, cinnarizine, Ph.Eur., RS
Organic reference standard, cinnarizine, Ph.Eur., RS
Catalog # EDQMC2180000
Supplier:  EDQM
CAS Number:  
Organic reference standard, cinnarizine, Ph.Eur., RS
Catalog # EDQMC2180000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    298-57-7
  • Description:
    Cinnarizine
  • Formula:
    C₂₆H₂₈N₂
  • Storage temperature:
    Ambient
  • Description:
    Cinnarizine

Specifications

About this item

  • Pharmaceutical industry applications