Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Chloramphenicol, Ph.Eur., RS
Organic reference standard, Chloramphenicol, Ph.Eur., RS
Catalog # EDQMC1200000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Chloramphenicol, Ph.Eur., RS
Catalog # EDQMC1200000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    500 mg
  • Standards:
    Reference standard
  • CAS No.:
    56-75-7
  • Description:
    Chloramphenicol
  • Formula:
    C₁₁H₁₂Cl₂N₂O₅
  • Storage temperature:
    Ambient
  • Description:
    Chloramphenicol

Specifications

About this item

  • Solubility in water at 25°C: 2,5 g/l
  • Pharmaceutical industry applications