Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Calcium pantothenate, Ph.Eur., RS
Organic reference standard, Calcium pantothenate, Ph.Eur., RS
Catalog # EDQMC0400000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Calcium pantothenate, Ph.Eur., RS
Catalog # EDQMC0400000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    137-08-6
  • Description:
    Calcium pantothenate
  • Formula:
    C₁₈H₃₂CaN₂O₁₀
  • Storage temperature:
    Ambient
  • Description:
    Calcium pantothenate

Specifications

About this item

  • Solubility in water at 25 °C: 350 g/l
  • Pharmaceutical industry applications