Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Butylhydroxytoluene, Ph.Eur., RS
Organic reference standard, Butylhydroxytoluene, Ph.Eur., RS
Catalog # EDQMB1215000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Butylhydroxytoluene, Ph.Eur., RS
Catalog # EDQMB1215000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    120 mg
  • Standards:
    Reference standard
  • CAS No.:
    128-37-0
  • Description:
    Butylhydroxytoluene
  • Formula:
    C₁₅H₂₄O
  • Storage temperature:
    Ambient
  • Description:
    Butylhydroxytoluene

Specifications

About this item

  • Plastic additive 07